Vanillyl butyl ether ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4-(Butoxymethyl)-2-methoxyphenol

V0161-25G V0161-5G
TCV0161-25GEA 77 USD
TCV0161-25G TCV0161-5G
Vanillyl butyl ether ≥98.0% (by GC)
Vanillyl butyl ether
Formula: C₁₂H₁₈O₃
Boiling Pt: 241 °C
MDL Number: MFCD00238529
CAS Number: 82654-98-6

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR