Oxymetazoline hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: SOxymetazoline hydrochloride, 6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-2,4-dimethylphenol hydrochloride, Afrin hydrochloride

O0520-25G O0520-5G
TCO0520-25GEA 489 USD
TCO0520-25G TCO0520-5G
Oxymetazoline hydrochloride ≥98.0% (by HPLC, titration analysis)
Oxymetazoline hydrochloride
Formula: C₁₆H₂₄N₂O·HCl
MW: 296.839 g/mol
Melting Pt: 303 °C
MDL Number: MFCD00058147
CAS Number: 2315-02-8
EINECS: 219-015-0
UN: 2811
ADR: 6.1 I
Merck Index: 14,06966

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR