(-)-Huperzine A ≥98.0% (by HPLC)

Supplier: TCI America
Danger

Synonyms: (−)-Selagine, (5R,9R,E)-5-Amino-11-ethylidene-7-methyl-5,6,9,10-tetrahydro-5,9-methanocycloocta[b]pyridin-2(1H)-one

H1700-100MG H1700-10MG
TCH1700-100MGEA 189 USD
TCH1700-100MG TCH1700-10MG
(-)-Huperzine A ≥98.0% (by HPLC)
(-)-Huperzine A
Formula: C₁₅H₁₈N₂O
MW: 242.32 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00171439
CAS Number: 102518-79-6
UN: 1544
ADR: 6.1,II
Merck Index: 14,04755

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 230.0 to 234.0 deg-C
Specific rotation [a]20/D -144 to -155 deg (C=0.5, MeOH)
Water max. 3.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR