Furan-2,5-dicarbonyl dichloride ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 2,5-Furandicarbonyl dichloride

F1022-1G F1022-5G
TCF1022-1GEA 114 USD
TCF1022-1G TCF1022-5G
Furan-2,5-dicarbonyl dichloride ≥98.0% (by GC)
Furan-2,5-dicarbonyl dichloride
Formula: C₆H₂Cl₂O₃
MW: 192.99 g/mol
Boiling Pt: 245 °C
Storage Temperature: Refrigerator
MDL Number: MFCD28044847
CAS Number: 10375-34-5
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 97.0 %
Melting point 76.0 to 80.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR