2,4-Dichloro-6-phenylpyrimidine ≥98.0% (by GC)

Supplier: TCI America
Warning

D5505-1G D5505-5G
TCD5505-1GEA 142 USD
TCD5505-1G TCD5505-5G
2,4-Dichloro-6-phenylpyrimidine ≥98.0% (by GC)
2,4-Dichloro-6-phenylpyrimidine
Formula: C₁₀H₆Cl₂N₂
MDL Number: MFCD00234591
CAS Number: 26032-72-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 85.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR