1-Ethynyl-3,5-difluorobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 3,5-Difluorophenylacetylene

E1352-1G E1352-5G
TCE1352-1GEA 70 USD
TCE1352-1G TCE1352-5G
1-Ethynyl-3,5-difluorobenzene ≥98.0% (by GC)
1-Ethynyl-3,5-difluorobenzene
Formula: C₈H₄F₂
Flash Pt: 33 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039982
CAS Number: 151361-87-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR