4-Ethynylbenzyl alcohol ≥98.0% (by GC)

Supplier: TCI America

E1377-1G E1377-5G
TCE1377-1GEA 73 USD
TCE1377-1G TCE1377-5G
4-Ethynylbenzyl alcohol ≥98.0% (by GC)
4-Ethynylbenzyl alcohol
Formula: C₉H₈O
Boiling Pt: 66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05664205
CAS Number: 10602-04-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR