Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate ≥98.0% (by GC)

Supplier: TCI America
Warning

E1247-1G E1247-5G
TCE1247-1GEA 65 USD
TCE1247-1G TCE1247-5G
Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate ≥98.0% (by GC)
Ethyl-2-amino-4-hydroxypyrimidine-5-carboxylate
Formula: C₇H₉N₃O₃
Melting Pt: 280 °C
MDL Number: MFCD00039708
CAS Number: 15400-53-0
EINECS: 239-418-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR