Ethyl-4-(4-methylphenyl)-4-oxobutyrate ≥95.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 4-(4-Methylphenyl)-4-oxobutanoate, 4-Oxo-4-(p-tolyl)butyric acid ethyl ester, Ethyl 4-oxo-4-(p-tolyl)butanoate, 4-(4-Methylphenyl)-4-oxobutanoic acid ethyl ester, Ethyl 4-Oxo-4-(p-tolyl)butyrate

E1195-1G E1195-5G
TCE1195-1GEA 213 USD
TCE1195-1G TCE1195-5G
Ethyl-4-(4-methylphenyl)-4-oxobutyrate ≥95.0% (by GC)
Ethyl-4-(4-methylphenyl)-4-oxobutyrate
Formula: C₁₃H₁₆O₃
MW: 220.27 g/mol
Boiling Pt: 178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00266564
CAS Number: 6942-61-6

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 95.0 %
Melting point 41.0 to 45.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR