4,6-Di(tert-butyl)benzene-1,3-diol ≥98.0% (by GC)

Supplier: TCI America
Warning

D5459-25G D5459-5G
TCD5459-25GEA 264 USD
TCD5459-25G TCD5459-5G
4,6-Di(tert-butyl)benzene-1,3-diol ≥98.0% (by GC)
4,6-Di(tert-butyl)benzene-1,3-diol
Formula: C₁₄H₂₂O₂
MDL Number: MFCD00044513
CAS Number: 5374-06-1
EINECS: 226-366-3

Order Now

Specification Test Results

Appearance White to Gray to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 121.0 to 125.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR