2,6-Dibromobenzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

D5485-1G D5485-5G
TCD5485-1GEA 77 USD
TCD5485-1G TCD5485-5G
2,6-Dibromobenzoic acid ≥98.0% (by GC, titration analysis)
2,6-Dibromobenzoic acid
Formula: C₇H₄Br₂O₂
Boiling Pt: 335 °C
MDL Number: MFCD00234254
CAS Number: 601-84-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 150.0 to 154.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR