1-(Ethylamino)propan-2-ol ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Danger

E1244-25ML E1244-5ML
TCE1244-25MLEA 153 USD
TCE1244-25ML TCE1244-5ML
1-(Ethylamino)propan-2-ol ≥97.0% (by titrimetric analysis)
1-(Ethylamino)propan-2-ol
Formula: C₅H₁₃NO
Boiling Pt: 160 °C
Melting Pt: 14 °C
MDL Number: MFCD00082193
CAS Number: 40171-86-6
EINECS: 254-821-6
UN: 2735
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR