(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

Synonyms: (S)-2-[(S)-4-Methyl-2,5-dioxooxazolidin-3-yl]-4-phenylbutanoic acid ethyl ester, Ethyl (S)-2-[(S)-4-Methyl-2,5-dioxooxazolidin-3-yl]-4-phenylbutanoate, Ethyl (S)-2-[(S)-4-Methyl-2,5-dioxo-3-oxazolidinyl]-4-phenylbutanoate, N-[1-(S)-Ethoxycarbonyl-3-phenylpropyl]-L-alanine-N-carboxyanhydride

E1423-1G E1423-5G
TCE1423-1GEA 21 USD
TCE1423-1G TCE1423-5G
(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate ≥98.0% (by HPLC, titration analysis)
(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate
Formula: C₁₆H₁₉NO₅
MW: 305.33 g/mol
Boiling Pt: 345 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00272296
CAS Number: 84793-24-8
EINECS: 430-360-8

Order Now

Specification Test Results

Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C
Specific rotation [a]20/D 34.0 to 38.0 deg (C=1, CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR