Desvenlafaxine ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4-(2-(Dimethylamino)-1-(1-hydroxycyclohexyl)-ethyl)phenol, O-Desmethylvenlafaxine, 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, Desvenlafaxine

D5598-1G D5598-5G
TCD5598-1GEA 108 USD
TCD5598-1G TCD5598-5G
Desvenlafaxine ≥98.0% (by HPLC)
Desvenlafaxine
Formula: C₁₆H₂₅NO₂
MW: 263.38 g/mol
MDL Number: MFCD00871934
CAS Number: 93413-62-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 220.0 to 224.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR