N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Gefitinib

G0546-1G G0546-5G
TCG0546-1GEA 84 USD
TCG0546-1G TCG0546-5G
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine ≥98.0% (by HPLC, titration analysis)
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine
Formula: C₂₂H₂₄ClFN₄O₃
MW: 446.91 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04307832
CAS Number: 184475-35-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 194.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR