Arabinofuranosyl-2-fluoroadenine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Fluoroadenine Arabinosid, (2R,3S,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 9-beta-D-Arabinofuranosyl-2-fluoro-9H-purin-6-amine, 9-beta-D-Arabinofuranosyl-2-fluoroadenine, Fludarabine

F0658-1G F0658-250MG
TCF0658-1GEA 92 USD
TCF0658-1G TCF0658-250MG
Arabinofuranosyl-2-fluoroadenine ≥98.0% (by HPLC, titration analysis)
Arabinofuranosyl-2-fluoroadenine
Formula: C₁₀H₁₂FN₅O₄
MW: 285.24 g/mol
Melting Pt: 260 °C
MDL Number: MFCD00132942
CAS Number: 21679-14-1
EINECS: 244-525-5
Merck Index: 14,04126

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +16.0 to +20.0 deg (C=0.1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR