Lafutidine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: (+)-(Z)-2-((Furan-2-ylmethyl)sulphinyl)-N-(4-((4-(piperidin-1-ylmethyl)pyridin-2-yl)oxy)but-2-en-1-yl)acetamide, FRG-8813, (Z)-2-((Furan-2-ylmethyl)sulphinyl)-N-(4-((3-(piperidin-1-ylmethyl)pyridin-2-yl)oxy)but-2-en-1-yl)acetamide, (+)-Lafutidine

L0341-1G L0341-250MG L0341-25MG
TCL0341-1GEA 267 USD
TCL0341-1G TCL0341-250MG TCL0341-25MG
Lafutidine ≥98.0% (by HPLC, titration analysis)
Lafutidine
Formula: C₂₂H₂₉N₃O₄S
MW: 431.55 g/mol
MDL Number: MFCD21607521
CAS Number: 118288-08-7
Merck Index: 14,05347

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 97.0 to 101.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR