4-Phenyl-2-pyrrolidone ≥98.0% (by GC)

Supplier: TCI America
Warning

P2727-1G P2727-5G
TCP2727-1GEA 85 USD
TCP2727-1G TCP2727-5G
4-Phenyl-2-pyrrolidone ≥98.0% (by GC)
4-Phenyl-2-pyrrolidone
Formula: C₁₀H₁₁NO
Boiling Pt: 178 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01687226
CAS Number: 1198-97-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 74.0 to 78.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR