Methyl-4-aminobutanoate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Supplier: TCI America
Synonyms:
H-γ-Abu-OMe.HCl
M3154-25G
M3154-5G
TCM3154-25GEA
198
USD
TCM3154-25G
TCM3154-5G
Methyl-4-aminobutanoate hydrochloride ≥98.0% (by total nitrogen and titration analysis)
Methyl-4-aminobutanoate hydrochloride
Formula:
C₅H₁₁NO₂·HCl MW: 153.61 g/mol |
MDL Number:
MFCD00043270 CAS Number: 13031-60-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 98.0 % |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 122.0 to 126.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...