Methyl-4-chloro-4-oxobutanoate ≥95.0% (by GC)

Supplier: TCI America
Danger

M2850-25G M2850-5G
TCM2850-25GEA 163 USD
TCM2850-25G TCM2850-5G
Methyl-4-chloro-4-oxobutanoate ≥95.0% (by GC)
Methyl-4-chloro-4-oxobutanoate
Formula: C₅H₇ClO₃
Boiling Pt: 65 °C
Flash Pt: 74 °C
MDL Number: MFCD00000750
CAS Number: 1490-25-1
EINECS: 216-077-0
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR