1H,1H-Heptafluoro-n-butyl methacrylate ≥98.0% (by GC) stabilised

Supplier: TCI America
Warning

Synonyms: 2,2,3,3,4,4,4-Heptafluorobutyl Methacrylate, 1H,1h-heptafluorobutylmethacrylate, Methacrylic acid 2,2,3,3,4,4,4-Heptafluorobutyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

H1674-25G H1674-5G
TCH1674-25GEA 251 USD
TCH1674-25G TCH1674-5G
1H,1H-Heptafluoro-n-butyl methacrylate ≥98.0% (by GC) stabilised
1H,1H-Heptafluoro-n-butyl methacrylate
Formula: C₈H₇F₇O₂
MW: 268.13 g/mol
Flash Pt: 37 °C
MDL Number: MFCD00039251
CAS Number: 13695-31-3
EINECS: 237-217-7
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR