(3,3-Difluorocyclobutyl)methanol ≥98.0% (by GC)

Supplier: TCI America
Warning

D5582-1G D5582-200MG
TCD5582-1GEA 246 USD
TCD5582-1G TCD5582-200MG
(3,3-Difluorocyclobutyl)methanol ≥98.0% (by GC)
(3,3-Difluorocyclobutyl)methanol
Formula: C₅H₈F₂O
MDL Number: MFCD11877861
CAS Number: 681128-39-2
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR