2,5-Diphenylpyridine ≥98.0% (by GC)

Supplier: TCI America

D5456-1G D5456-5G
TCD5456-1GEA 113 USD
TCD5456-1G TCD5456-5G
2,5-Diphenylpyridine ≥98.0% (by GC)
2,5-Diphenylpyridine
Formula: C₁₇H₁₃N
Boiling Pt: 255 °C
MDL Number: MFCD00444961
CAS Number: 15827-72-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 171.0 to 175.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR