3-Nitro-2-pyridinethiol ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

N1149-1G N1149-5G
TCN1149-1GEA 58 USD
TCN1149-1G TCN1149-5G
3-Nitro-2-pyridinethiol ≥98.0% (by HPLC, titration analysis)
3-Nitro-2-pyridinethiol
Formula: C₅H₄N₂O₂S
MDL Number: MFCD00661369
CAS Number: 38240-29-8

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 172.0 to 176.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR