Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Methyl 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionate, 3-(3,5-Di-tert-butyl-4-hydroxyphenyl)propionic acid methyll ester, Methyl 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate

M1810-25G M1810-5G
TCM1810-25GEA 45 USD
TCM1810-25G TCM1810-5G
Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate ≥98.0% (by GC)
Methyl-3-[3,5-di(tert-butyl)-4-hydroxyphenyl]propanoate
Formula: C₁₈H₂₈O₃
MW: 292.42 g/mol
Boiling Pt: 130 °C
MDL Number: MFCD00210461
CAS Number: 6386-38-5
EINECS: 228-985-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 65.0 to 69.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR