1H-Indole-3-carbaldehyde oxime ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1H-Indole-3-carbaldehyde Oxime, 1H-Indole-3-carboxaldehyde Oxime

I1060-1G I1060-5G
TCI1060-1GEA 102 USD
TCI1060-1G TCI1060-5G
1H-Indole-3-carbaldehyde oxime ≥98.0% (by GC)
1H-Indole-3-carbaldehyde oxime
Formula: C₉H₈N₂O
MW: 160.18 g/mol
MDL Number: MFCD00022737
CAS Number: 2592-05-4

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR