3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
3-(4-Fluorophenyl)pyrazole-4-carboxaldehyde
F1118-1G
F1118-5G
TCF1118-1GEA
109
USD
TCF1118-1G
TCF1118-5G
3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
3-(4-Fluorophenyl)-1H-pyrazole-4-carbaldehyde
Formula:
C₁₀H₇FN₂O MW: 190.18 g/mol |
MDL Number:
MFCD01922131 CAS Number: 306936-57-2 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Acid Value | max. 2.0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...