4-Isopropylbenzylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

I1074-5ML I1074-1ML
TCI1074-5MLEA 281 USD
TCI1074-5ML TCI1074-1ML
4-Isopropylbenzylamine ≥98.0% (by GC, titration analysis)
4-Isopropylbenzylamine
Formula: C₁₀H₁₅N
Boiling Pt: 227 °C
MDL Number: MFCD00025580
CAS Number: 4395-73-7
UN: 2735
ADR: 8,III

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR