5-Hydroxy-1H-indazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
1H-Indazol-5-ol
H1636-1G
H1636-5G
TCH1636-1GEA
54
USD
TCH1636-1G
TCH1636-5G
5-Hydroxy-1H-indazole ≥98.0% (by GC)
5-Hydroxy-1H-indazole
Formula:
C₇H₆N₂O MW: 134.14 g/mol |
MDL Number:
MFCD01938124 CAS Number: 15579-15-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 187.0 to 191.0 deg-C |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...