Pyrimidine-2-sulfonyl fluoride ≥98.0% (by GC)

Supplier: TCI America
Danger

P2465-5G P2465-1G
TCP2465-5GEA 429 USD
TCP2465-5G TCP2465-1G
Pyrimidine-2-sulfonyl fluoride ≥98.0% (by GC)
Pyrimidine-2-sulfonyl fluoride
Formula: C₄H₃FN₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD09265503
CAS Number: 35762-87-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Purity (Neutralization back titration) min. 97.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR