Trimethyl(2-methylprop-2-en-1-yl)silane ≥95.0% (by GC)

Supplier: TCI America
Danger

M3010-25ML M3010-5ML
TCM3010-25MLEA 325 USD
TCM3010-25ML TCM3010-5ML
Trimethyl(2-methylprop-2-en-1-yl)silane ≥95.0% (by GC)
Trimethyl(2-methylprop-2-en-1-yl)silane
Formula: C₇H₁₆Si
Boiling Pt: 112 °C
Flash Pt: −6 °C
MDL Number: MFCD00054404
CAS Number: 18292-38-1
UN: 1993
ADR: 3,II

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR