1-Ethynyl-2,4-difluorobenzene ≥96.0% (by GC)

Supplier: TCI America
Warning

E1169-1G E1169-5G
TCE1169-1GEA 169 USD
TCE1169-1G TCE1169-5G
1-Ethynyl-2,4-difluorobenzene ≥96.0% (by GC)
1-Ethynyl-2,4-difluorobenzene
Formula: C₈H₄F₂
Storage Temperature: Refrigerator
MDL Number: MFCD02093726
CAS Number: 302912-34-1
UN: 1325
ADR: 4.1,III

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR