Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Fmoc-trans-4-Amc-OH, trans-4-(((((9H-fluoren-9-yl)methoxy)carbonyl)amino)methyl)cyclohexanecarboxylic acid, N-Fmoc-tranexamic acid, trans-4-[[[[(9H-Fluoren-9-yl)methoxy]carbonyl]amino]methyl]cyclohexanecarboxylic acid, trans-4-(Fmoc-aminomethyl)cyclohexanecarboxylic acid

F1215-1G F1215-5G
TCF1215-1GEA 29 USD
TCF1215-1G TCF1215-5G
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid ≥98.0% (by HPLC, titration analysis)
Fmoc-trans-4-(aminomethyl)cyclohexane-1-carboxylic acid
Formula: C₂₃H₂₅NO₄
MW: 379.46 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00273480
CAS Number: 167690-53-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 186.0 to 190.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR