3-(Perfluorohexyl)propanol-1 ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1H,1H,2H,2H,3H,3H-Tridecafluoro-1-nonanol, 3-(Perfluorohexyl)propanol

T3258-25G T3258-5G
TCT3258-25GEA 218 USD
TCT3258-25G TCT3258-5G
3-(Perfluorohexyl)propanol-1 ≥98.0% (by GC)
3-(Perfluorohexyl)propanol-1
Formula: C₉H₇F₁₃O
Boiling Pt: 86 °C
Melting Pt: −3 °C
MDL Number: MFCD00077578
CAS Number: 80806-68-4
UN: 1987
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR