Malonic acid dihydrazide ≥93.0% (by titrimetric analysis)

Supplier: TCI America
Warning

M3206-25G M3206-5G
TCM3206-25GEA 135 USD
TCM3206-25G TCM3206-5G
Malonic acid dihydrazide ≥93.0% (by titrimetric analysis)
Malonic acid dihydrazide
Formula: C₃H₈N₄O₂
MW: 132.12 g/mol
MDL Number: MFCD00041268
CAS Number: 3815-86-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 93.0 %
Melting point 156.0 to 160.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR