2-Ethoxy-4-formylphenyl acetate ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Ethylvanillin acetate

E1415-25G E1415-5G
TCE1415-25GEA 94 USD
TCE1415-25G TCE1415-5G
2-Ethoxy-4-formylphenyl acetate ≥98.0% (by GC)
2-Ethoxy-4-formylphenyl acetate
Formula: C₁₁H₁₂O₄
MW: 208.22 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01666413
CAS Number: 72207-94-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 43.0 to 47.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR