6H-Indolo[2,3-b]quinoxaline ≥98.0% (by GC, titration analysis)

Supplier: TCI America

I1098-1G I1098-200MG
TCI1098-1GEA 204 USD
TCI1098-1G TCI1098-200MG
6H-Indolo[2,3-b]quinoxaline ≥98.0% (by GC, titration analysis)
6H-Indolo[2,3-b]quinoxaline
Formula: C₁₄H₉N₃
MDL Number: MFCD00068754
CAS Number: 84055-81-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR