4-(Trifluoromethyl)cyclohexylamine cis- and trans-mixture ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 4-(Trifluoromethyl)cyclohexanamine

T3613-1G T3613-5G
TCT3613-1GEA 96 USD
TCT3613-1G TCT3613-5G
4-(Trifluoromethyl)cyclohexylamine cis- and trans-mixture ≥98.0% (by GC)
4-(Trifluoromethyl)cyclohexylamine
Formula: C₇H₁₂F₃N
MW: 167.18 g/mol
Boiling Pt: 71 °C
MDL Number: MFCD09040573
CAS Number: 58665-70-6
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Colourless to Light yellow clear liquid
Purity (GC) min. 98.0 % (total of isomers)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR