Perfluoro-15-crown-5-ether 98,0 GC_ASSAY_METHOD

Supplier: TCI America
Warning

Synonyms: Icosafluoro-15-crown 5-Ether, Icosafluoro-1,4,7,10,13-pentaoxacyclopentadecane, Perfluoro-1,4,7,10,13-pentaoxacyclopentadecane, Perfluoro-15-crown 5-Ether

I1043-1G I1043-5G
TCI1043-1GEA 82 USD
TCI1043-1G TCI1043-5G
Perfluoro-15-crown-5-ether 98,0 GC_ASSAY_METHOD
Perfluoro-15-crown-5-ether
Formula: C₁₀F₂₀O₅
MW: 580.08 g/mol
Boiling Pt: 146 °C
Melting Pt: –12 °C
MDL Number: MFCD01320747
CAS Number: 97571-69-2

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR