Ethyl-2,4,5-trifluorobenzoylacetate ≥97.0% (by GC)

Supplier: TCI America

E1425-1G E1425-5G
TCE1425-1GEA 27 USD
TCE1425-1G TCE1425-5G
Ethyl-2,4,5-trifluorobenzoylacetate ≥97.0% (by GC)
Ethyl-2,4,5-trifluorobenzoylacetate
Formula: C₁₁H₉F₃O₃
Boiling Pt: 92 °C
MDL Number: MFCD00792431
CAS Number: 98349-24-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 65.0 to 69.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR