3-Phenylumbelliferone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 7-Hydroxy-3-phenyl-2H-1-benzopyran-2-one, 7-Hydroxy-3-phenylcoumarin, 7-Hydroxy-3-phenyl-2H-chromen-2-one

P2540-1G P2540-5G
TCP2540-1GEA 121 USD
TCP2540-1G TCP2540-5G
3-Phenylumbelliferone ≥98.0% (by GC)
3-Phenylumbelliferone
Formula: C₁₅H₁₀O₃
MW: 238.242 g/mol
MDL Number: MFCD00037574
CAS Number: 6468-96-8

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 211.0 to 215.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR