Sclareol ≥96.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (1R,2R,4aS,8aS)-1-((R)-3-Hydroxy-3-methylpent-4-en-1-yl)-2,5,5,8a-tetramethyldecahydronaphthalen-2-ol

S0916-1G S0916-5G
TCS0916-1GEA 23 USD
TCS0916-1G TCS0916-5G
Sclareol ≥96.0% (by GC)
Sclareol
Formula: C₂₀H₃₆O₂
MW: 308.51 g/mol
Boiling Pt: 189 °C
MDL Number: MFCD00869558
CAS Number: 515-03-7
EINECS: 208-194-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 96.0 %
Melting point 99.0 to 103.0 deg-C
Specific rotation [a]20/D -5.0 to -3.0 deg (C=1,CHCl3)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR