Fmoc-L-Tyr(Me)-OH ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: Fmoc-Tyr(Me)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-O-methyl-L-tyrosine, N-Fmoc-4-methoxy-L-phenylalanine, N-Fmoc-O-methyl-L-tyrosine, Fmoc-Phe(4-OMe)-OH, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-methoxy-L-phenylalanine

F1038-1G F1038-5G
TCF1038-1GEA 83 USD
TCF1038-1G TCF1038-5G
Fmoc-L-Tyr(Me)-OH ≥98.0% (by HPLC, titration analysis)
Fmoc-L-Tyr(Me)-OH
Formula: C₂₅H₂₃NO₅
MW: 417.47 g/mol
MDL Number: MFCD00153368
CAS Number: 77128-72-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 159.0 to 163.0 deg-C
Specific rotation [a]20/D -26.0 to -30.0 deg (C=1,DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR