Sulindac ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

U0099-10G U0099-50G
TCU0099-10GEA 136 USD
TCU0099-10G TCU0099-50G
Sulindac ≥98.0% (by HPLC, titration analysis)
Sulindac
Formula: C₂₀H₁₇FO₃S
MDL Number: MFCD00599589
CAS Number: 38194-50-2
EINECS: 253-819-2
UN: 2811
ADR: 6.1,III
Merck Index: 14,08982

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 182.0 to 186.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR