Methyl-4-trifluoromethylbenzoylacetate ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
Methyl 3-Oxo-3-[4-(trifluoromethyl)phenyl]propionoate, 3-Oxo-3-[4-(trifluoromethyl)phenyl]propionic acid methyll ester, Methyl 4-(Trifluoromethyl)benzoylacetate, 4-(Trifluoromethyl)benzoylacetic acid methyll ester, Methyl 3-oxo-3-(4-(trifluoromethyl)phenyl)propanoate
M2966-1G
M2966-5G
TCM2966-1GEA
36
USD
TCM2966-1G
TCM2966-5G
Methyl-4-trifluoromethylbenzoylacetate ≥95.0% (by GC)
Methyl-4-trifluoromethylbenzoylacetate
Formula:
C₁₁H₉F₃O₃ MW: 246.185 g/mol |
MDL Number:
MFCD00216521 CAS Number: 212755-76-5 |
Specification Test Results
Appearance | Light yellow to Yellow to Orange powder to crystal |
Purity (GC) | min. 95.0 % |
Melting point | 58.0 to 62.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...