Phenyl-p-benzoquinone ≥96.0% (by GC)

Supplier: TCI America

Synonyms: 2-Phenyl-1,4-benzoquinone, [1,1'-Biphenyl]-2,5-dione

P2674-1G P2674-5G
TCP2674-1GEA 38 USD
TCP2674-1G TCP2674-5G
Phenyl-p-benzoquinone ≥96.0% (by GC)
Phenyl-p-benzoquinone
Formula: C₁₂H₈O₂
MW: 184.194 g/mol
MDL Number: MFCD00001599
CAS Number: 363-03-1
EINECS: 206-654-5

Order Now

Specification Test Results

Appearance White to Brown to Dark purple powder to crystal
Purity (GC) min. 96.0 %
Melting point 111.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR