(S)-1-(Pyridin-4-yl)ethanol ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

H1734-1G H1734-5G
TCH1734-1GEA 95 USD
TCH1734-1G TCH1734-5G
(S)-1-(Pyridin-4-yl)ethanol ≥97.0% (by GC, titration analysis)
(S)-1-(Pyridin-4-yl)ethanol
Formula: C₇H₉NO
MDL Number: MFCD00077866
CAS Number: 54656-96-1

Order Now

Specification Test Results

Appearance White to Light red to Green powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %
Purity (GC) min. 97.0 %
Optical purity (GC) min. 98.0 ee%
Melting point 67.0 to 71.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR