3-(Trifluoromethyl)styrene ≥97.0% (by GC) stabilized

Supplier: TCI America
Warning

Synonyms: 1-(Trifluoromethyl)-3-vinylbenzene

Stabilization: Stabilised with hydroquinone (HQ)

T3354-1G T3354-5G
TCT3354-1GEA 129 USD
TCT3354-1G TCT3354-5G
3-(Trifluoromethyl)styrene ≥97.0% (by GC) stabilized
3-(Trifluoromethyl)styrene
Formula: C₉H₇F₃
MW: 172.15 g/mol
Boiling Pt: 65 °C
Flash Pt: 42 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00075481
CAS Number: 402-24-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR