8-hydroxyquinoline-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 8-Hydroxyquinoline-2-carboxylic acid

H1658-1G H1658-5G
TCH1658-1GEA 124 USD
TCH1658-1G TCH1658-5G
8-hydroxyquinoline-2-carboxylic acid ≥98.0% (by GC, titration analysis)
8-hydroxyquinoline-2-carboxylic acid
Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
MDL Number: MFCD00168972
CAS Number: 1571-30-8

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR