1-Hydroxy-1-cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-hydroxycyclopropane-1-carboxylic acid, 1-Hydroxycyclopropanecarboxylic acid

H1664-1G H1664-200MG
TCH1664-1GEA 91 USD
TCH1664-1G TCH1664-200MG
1-Hydroxy-1-cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-Hydroxy-1-cyclopropanecarboxylic acid
Formula: C₄H₆O₃
MW: 102.089 g/mol
MDL Number: MFCD00010797
CAS Number: 17994-25-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 107.0 to 111.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR